Assay Details
| Gene Symbol: Alcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009655.2 | NP_033785.1 | 13 - 14 | 2078 | 75 | |
| GenBank mRNA | AB003026.1 | - | 13 - 14 | 1740 | 75 | |
| AK030851.1 | - | 13 - 14 | 2079 | 75 | ||
| AK031391.1 | - | 3154 | 75 | |||
| BC027280.1 | - | 13 - 14 | 1735 | 3501215 | 75 | |
| L25274.1 | - | 8 - 9 | 870 | 75 | ||
| U95030.1 | - | 13 - 14 | 1546 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alcam
Genomic Map