Assay Details
| Gene Symbol: Alcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009655.2 | NP_033785.1 | 7 - 8 | 1385 | 102 | |
| GenBank mRNA | AB003026.1 | - | 7 - 8 | 1047 | 102 | |
| AK030851.1 | - | 7 - 8 | 1386 | 102 | ||
| AK031391.1 | - | 2461 | 102 | |||
| BC027280.1 | - | 7 - 8 | 1042 | 3501215 | 102 | |
| L25274.1 | - | 2 - 3 | 177 | 102 | ||
| U95030.1 | - | 7 - 8 | 853 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alcam
Genomic Map