Assay Details
| Gene Symbol: Alcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009655.2 | NP_033785.1 | 8 - 9 | 1523 | 62 | |
| GenBank mRNA | AB003026.1 | - | 8 - 9 | 1185 | 62 | |
| AK030851.1 | - | 8 - 9 | 1524 | 62 | ||
| AK031391.1 | - | 2599 | 62 | |||
| BC027280.1 | - | 8 - 9 | 1180 | 3501215 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alcam
Genomic Map