Assay Details
| Gene Symbol: Rhbdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291818.1 | NP_001278747.1 | 10 - 11 | 1430 | 75 | |
| NM_010117.2 | NP_034247.2 | 10 - 11 | 1549 | 75 | ||
| GenBank mRNA | AK144437.1 | - | 10 - 11 | 1418 | 75 | |
| AK198332.1 | - | 1 - 2 | 39 | 75 | ||
| BC023469.1 | - | 5 - 6 | 697 | 75 | ||
| BC027346.1 | - | 10 - 11 | 1494 | 4011421 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf1
Genomic Map