Assay Details
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010140.3 | NP_034270.1 | 7 - 8 | 1699 | 67 | |
GenBank mRNA | AK045219.1 | - | 7 - 8 | 1700 | 67 | |
AK084921.1 | - | 7 - 8 | 1679 | 67 | ||
AK132335.1 | - | 7 - 8 | 1648 | 67 | ||
BC053479.1 | - | 3 - 4 | 591 | 67 | ||
BC060164.1 | - | 7 - 8 | 1676 | 67 | ||
BC093483.1 | - | 1700 | 67 | |||
BC138641.1 | - | 7 - 8 | 1622 | 8861661 | 67 | |
BC138643.1 | - | 7 - 8 | 1622 | 8861663 | 67 | |
BC145349.1 | - | 7 - 8 | 1619 | 9053905 | 67 | |
M68513.1 | - | 7 - 8 | 1674 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
