Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100022612.mm.1 |
NM_025740.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022612.mm.1 |
NM_025740.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022612.mm.1 |
TC1100002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022609.mm.1 |
NM_025740.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022609.mm.1 |
NM_025740.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022609.mm.1 |
TC1100002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022611.mm.1 |
NM_025740.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022611.mm.1 |
NM_025740.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022611.mm.1 |
TC1100002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022614.mm.1 |
NM_025740.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022614.mm.1 |
NM_025740.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022614.mm.1 |
TC1100002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022613.mm.1 |
NM_025740.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022613.mm.1 |
NM_025740.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022613.mm.1 |
TC1100002416.mm.1 |
MTA TC |