Assay Details
| Gene Symbol: Cfap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025740.3 | NP_080016.1 | 7 - 8 | 784 | 117 | |
| GenBank mRNA | AK016479.1 | - | 7 - 8 | 819 | 117 | |
| AK075858.1 | - | 7 - 8 | 785 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cfap36
Genomic Map