Assay Details
| Gene Symbol: Mrpl52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026851.2 | NP_081127.1 | 2 - 3 | 106 | 118 | |
| GenBank mRNA | AK004194.1 | - | 2 - 3 | 107 | 118 | |
| AK150806.1 | - | 2 - 3 | 106 | 118 | ||
| AK151615.1 | - | 2 - 3 | 106 | 118 | ||
| AK152876.1 | - | 2 - 3 | 93 | 118 | ||
| AK164629.1 | - | 2 - 3 | 104 | 118 | ||
| BC060043.1 | - | 1 - 2 | 116 | 5683366 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060043.1 transcripts.
Set Membership: |
|

Mrpl52
Genomic Map