Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700023652.mm.1 |
TC0700004643.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023652.mm.1 |
NM_027470.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023652.mm.1 |
NM_027470.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023659.mm.1 |
TC0700004643.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023659.mm.1 |
NM_027470.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023659.mm.1 |
NM_027470.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023654.mm.1 |
TC0700004643.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023654.mm.1 |
NM_027470.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023654.mm.1 |
NM_027470.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039174.mm.1 |
NM_027470.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039174.mm.1 |
NM_027470.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039174.mm.1 |
TC0700004643.mm.1 |
MTA TC |