Assay Details
Gene Symbol: Pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027470.3 | NP_081746.1 | 2 - 3 | 223 | 94 | |
GenBank mRNA | AK129298.2 | - | 2 - 3 | 219 | 94 | |
BB839524.1 | - | 2 - 3 | 197 | 94 | ||
BC048238.1 | - | 2 - 3 | 185 | 6475114 | 94 | |
BY197606.1 | - | 2 - 3 | 223 | 94 | ||
CF172418.1 | - | 1 - 2 | 42 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC048238.1, NM_027470.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
