Assay Details
| Gene Symbol: Pak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027470.3 | NP_081746.1 | 8 - 9 | 1782 | 68 | |
| GenBank mRNA | AK017713.1 | - | 3 - 4 | 406 | 68 | |
| AK088512.1 | - | 8 - 9 | 1759 | 68 | ||
| AK129298.2 | - | 8 - 9 | 1778 | 68 | ||
| AY217016.1 | - | 6 - 7 | 1497 | 68 | ||
| BC030389.1 | - | 5 - 6 | 940 | 68 | ||
| BC048238.1 | - | 8 - 9 | 1744 | 6475114 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pak4
Genomic Map