Assay Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033560.3 | NP_291038.2 | 9 - 10 | 1300 | 72 | |
| GenBank mRNA | AK016385.1 | - | 9 - 10 | 1283 | 72 | |
| AK157507.1 | - | 9 - 10 | 1267 | 72 | ||
| AK171055.1 | - | 9 - 10 | 1289 | 72 | ||
| AK218075.1 | - | 4 - 5 | 301 | 72 | ||
| BC039290.1 | - | 9 - 10 | 1256 | 5066508 | 72 | |
| BC110396.1 | - | 9 - 10 | 1215 | 5253683 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37a
Genomic Map