Assay Details
Gene Symbol: Zmym5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036513.1 | - | 5 - 6 | 1075 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29624840 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29624841 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29624854 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041050 | AK165863.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041050 | NM_144842.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2463930 | AK155333.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400065 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719289 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550726 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494775 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365435 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376894 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848325 | AK085200.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848325 | AK085233.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848325 | XM_001480114.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848325 | XM_001480324.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406014 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18705 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231597 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272085 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526983 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14544 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160113 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166794 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113440 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2463930 | AK155333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041050 | AK165863.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041050 | NM_144842.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624874 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624835 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29624834 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08121967 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166794 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113440 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08121931 | - | - |
Whole Mouse Genome G4122A | A_51_P225981 | AK165863.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225981 | BC027380.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225981 | AK156501.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225981 | AK163025.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225981 | AK165292.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P277695 | AK165863.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P277695 | AK156501.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P277695 | AK163025.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P277695 | AK165292.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P277695 | NM_144842.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288288 | CN534727.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288288 | BC027380.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288288 | AK156501.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288288 | AK036513.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P288288 | AK155333.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P179211 | BC058644.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106027 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166794 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113440 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11293674 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29624841 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29624840 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29624854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29624872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624883 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624860 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624784 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11480101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11372645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624811 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11482896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624886 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624835 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11556357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11480105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624795 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11368077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11200585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624874 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11482898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624891 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624864 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624798 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624836 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29624830 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10420352 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420351 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420350 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420356 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420353 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420349 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420354 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420355 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1442007_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1445543_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425577_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10420348 | - | - |
Mouse Genome 430 2.0 Array | 1425577_at | - | - |
Mouse Genome 430 2.0 Array | 1425577_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1442007_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445543_at | - | - |
Mouse Genome 430 2.0 Array | 1445543_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1442007_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 219105_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 219105_at | - | - |
Mouse Expression Set 430 | 1425577_PM_at | - | - |
Mouse Expression Set 430 | 1425577_at | - | - |
Mouse Expression Set 430 | 1445543_PM_at | - | - |
Mouse Expression Set 430 | 1442007_at | - | - |
Mouse Expression Set 430 | 1442007_PM_at | - | - |
Mouse Expression Set 430 | 1445543_at | - | - |
Murine Genome U74v2 Set | 135351_PM_i_at | - | - |
Murine Genome U74v2 Set | 135352_r_at | - | - |
Murine Genome U74v2 Set | 166543_r_at | - | - |
Murine Genome U74v2 Set | 135351_i_at | - | - |
Murine Genome U74v2 Set | 166543_PM_r_at | - | - |
Murine Genome U74v2 Set | 135352_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1425577_at | - | - |
Mouse Genome 430A 2.0 Array | 1425577_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425577_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425577_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442007_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445543_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445543_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442007_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6130072 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006130072 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231971 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1231971 | - | - |
mouse-6 v1.1 (Illumina) | scl0219105.1_166-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6130072 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0219105.1_166-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130072 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1231971 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231971 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0219105.1_166-S | - | - |
