Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100017704.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017704.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017702.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017702.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017703.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017703.mm.1 |
TC0100002204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017705.mm.1 |
NM_173870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017705.mm.1 |
TC0100002204.mm.1 |
MTA TC |