Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_001253831.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_001253831.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_001253834.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_001253834.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_001253836.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_001253836.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_175025.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
NM_175025.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024684.mm.1 |
TC0900002991.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_001253831.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_001253831.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_001253834.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_001253834.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_001253836.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_001253836.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_175025.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
NM_175025.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024685.mm.1 |
TC0900002991.mm.1 |
MTA TC |