Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175214.3 | NP_780423.2 | 11 - 12 | 2756 | 96 | |
| GenBank mRNA | AK032912.1 | - | 4 - 5 | 611 | 96 | |
| BC116646.1 | - | 9 - 10 | 1595 | 40086396 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif27
Genomic Map