Assay Details
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175563.5 | NP_780772.1 | 3 - 4 | 353 | 91 | |
GenBank mRNA | AK040348.1 | - | 3 - 4 | 331 | 91 | |
AK047463.1 | - | 3 - 4 | 331 | 91 | ||
AK083651.1 | - | 3 - 4 | 348 | 91 | ||
BC023918.1 | - | 3 - 4 | 363 | 5323297 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
