Assay Details
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043739.1 | - | 1 - 2 | 478 | 62 | |
| AK047449.1 | - | 1 - 2 | 468 | 62 | ||
| AK049811.1 | - | 1 - 2 | 412 | 62 | ||
More Information
Set Membership: |
|

Sybu
Genomic Map