Assay Details
Gene Symbol: Crb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177638.4 | NP_808306.1 | 3 - 4 | 330 | 93 | |
GenBank mRNA | AK077528.1 | - | 3 - 4 | 262 | 93 | |
AK167279.1 | - | 3 - 4 | 260 | 93 | ||
BC024462.1 | - | 3 - 4 | 330 | 4975562 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259848 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259850 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259849 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259845 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259846 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137979 | AK077528.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137979 | AK167279.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137979 | NM_177638.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | AK077528.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | AK167279.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | BC024462.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | AK077528.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | AK167279.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328089 | BC024462.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46046 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619594 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871748 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171826 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121348 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157246 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137979 | AK077528.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137979 | AK167279.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137979 | NM_177638.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | AK077528.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | AK167279.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | BC024462.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | AK077528.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | AK167279.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328089 | BC024462.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | AK077528.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | AK167279.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | BC024462.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | AK077528.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | AK167279.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328089 | BC024462.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259845 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259857 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171826 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121348 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157246 | - | - |
Whole Mouse Genome G4122A | A_51_P328089 | AK077528.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328089 | AK167279.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328089 | BC024462.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328089 | AK077528.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328089 | AK167279.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328089 | NM_177638.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328089 | BC024462.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130345 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121348 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157246 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171826 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13492415 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259845 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259850 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259846 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259849 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259839 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259859 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13492414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13081242 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13492412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259860 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259844 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259853 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259842 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259840 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31259850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13192156 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10446226 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10446228 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10446225 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10446227 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434851_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424809_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455115_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10446224 | - | - |
Mouse Genome 430 2.0 Array | 1455115_a_at | - | - |
Mouse Genome 430 2.0 Array | 1424809_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1434851_s_at | - | - |
Mouse Genome 430 2.0 Array | 1455115_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1424809_at | - | - |
Mouse Genome 430 2.0 Array | 1434851_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224912_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224912_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700009874.mm.1 | NM_177638.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009874.mm.1 | NM_177638.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009874.mm.1 | TC1700001023.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700009876.mm.1 | NM_177638.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009876.mm.1 | NM_177638.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009876.mm.1 | TC1700001023.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700009873.mm.1 | NM_177638.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009873.mm.1 | NM_177638.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009873.mm.1 | TC1700001023.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700009875.mm.1 | NM_177638.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009875.mm.1 | NM_177638.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009875.mm.1 | TC1700001023.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700009877.mm.1 | NM_177638.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009877.mm.1 | NM_177638.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700009877.mm.1 | TC1700001023.mm.1 | MTA TC |
Mouse Expression Set 430 | 1434851_PM_s_at | - | - |
Mouse Expression Set 430 | 1424809_at | - | - |
Mouse Expression Set 430 | 1424809_PM_at | - | - |
Mouse Expression Set 430 | 1455115_PM_a_at | - | - |
Mouse Expression Set 430 | 1455115_a_at | - | - |
Mouse Expression Set 430 | 1434851_s_at | - | - |
Murine Genome U74v2 Set | 104584_f_at | - | - |
Murine Genome U74v2 Set | 104584_PM_f_at | - | - |
Murine Genome U74v2 Set | 104585_at | - | - |
Murine Genome U74v2 Set | 104585_PM_at | - | - |
Murine Genome U74v2 Set | 168426_f_at | - | - |
Murine Genome U74v2 Set | 168426_PM_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1424809_at | - | - |
Mouse Genome 430A 2.0 Array | 1455115_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1424809_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1434851_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1434851_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1455115_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455115_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455115_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424809_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434851_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434851_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424809_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 830412 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6940091 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6940091 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006940091 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755888 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2755888 | - | - |
mouse-6 v1.1 (Illumina) | scl50580.5_35-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6940091 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl50580.5_35-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940091 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2755888 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755888 | - | - |
