Assay Details
Gene Symbol: Nek5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177898.4 | NP_808566.2 | 10 - 11 | 971 | 70 | |
GenBank mRNA | AK032672.1 | - | 10 - 11 | 973 | 70 | |
AK165108.1 | - | 8 - 9 | 720 | 70 | ||
BC053516.1 | - | 10 - 11 | 959 | 6311513 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
