Assay Details
Gene Symbol: Nek5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177898.4 | NP_808566.2 | 11 - 12 | 1035 | 95 | |
GenBank mRNA | AK032672.1 | - | 11 - 12 | 1037 | 95 | |
AK165108.1 | - | 9 - 10 | 784 | 95 | ||
BC053516.1 | - | 11 - 12 | 1023 | 6311513 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
