Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800014698.mm.1 |
NM_177898.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014698.mm.1 |
NM_177898.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014698.mm.1 |
TC0800001861.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014699.mm.1 |
NM_177898.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014699.mm.1 |
NM_177898.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014699.mm.1 |
TC0800001861.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014700.mm.1 |
NM_177898.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014700.mm.1 |
NM_177898.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014700.mm.1 |
TC0800001861.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014701.mm.1 |
NM_177898.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014701.mm.1 |
NM_177898.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014701.mm.1 |
TC0800001861.mm.1 |
MTA TC |