Assay Details
Gene Symbol: Nek5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177898.4 | NP_808566.2 | 7 - 8 | 629 | 76 | |
GenBank mRNA | AK032672.1 | - | 7 - 8 | 631 | 76 | |
AK054168.1 | - | 7 - 8 | 643 | 76 | ||
AK165108.1 | - | 5 - 6 | 392 | 76 | ||
BC053516.1 | - | 7 - 8 | 617 | 6311513 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
