| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900015905.mm.1 |
NM_001253869.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015905.mm.1 |
NM_001253869.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015905.mm.1 |
TC0900001888.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253867.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253867.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253868.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253868.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253869.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253869.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253870.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_001253870.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_178577.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NM_178577.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NR_045606.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
NR_045606.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015900.mm.1 |
TC0900001888.mm.1 |
MTA TC |