Assay Details
| Gene Symbol: Tmem205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253867.1 | NP_001240796.1 | 3 - 4 | 449 | 73 | |
| NM_001253868.1 | NP_001240797.1 | 3 - 4 | 522 | 73 | ||
| NM_001253869.1 | NP_001240798.1 | 3 - 4 | 381 | 73 | ||
| NM_001253870.1 | NP_001240799.1 | 3 - 4 | 509 | 73 | ||
| NM_178577.5 | NP_848692.2 | 3 - 4 | 517 | 73 | ||
| GenBank mRNA | AB030201.1 | - | 3 - 4 | 352 | 73 | |
| AI573565.1 | - | 3 - 4 | 333 | 73 | ||
| BC010787.1 | - | 3 - 4 | 444 | 4211629 | 73 | |
| CA326029.1 | - | 3 - 4 | 381 | 73 | ||
| CD775257.1 | - | 1 - 2 | 73 | |||
| CN840781.1 | - | 3 - 4 | 437 | 73 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503654 | BC010787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503654 | NM_178577.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503654 | AB030201.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503654 | BC010787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503654 | NM_178577.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503654 | AB030201.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843174 | AK006242.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843174 | AK007056.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843174 | AK133216.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843174 | NM_026088.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116590 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135486 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503654 | BC010787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503654 | NM_178577.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503654 | AB030201.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503654 | BC010787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503654 | NM_178577.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503654 | AB030201.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349548 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06719972 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135486 | - | - |
| Whole Mouse Genome G4122A | A_51_P503654 | BC010787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P503654 | NM_178577.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P503654 | AB030201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P503654 | BC010787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P503654 | NM_178577.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P503654 | AB030201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P268269 | AK003909.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119381 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135486 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08070766 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08121032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08070768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07558553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07960868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08121026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26349545 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591657 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591656 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591655 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430421_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591653 | - | - |
| Mouse Genome 430 2.0 Array | 1430421_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430421_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235043_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235043_PM_at | - | - |
| Mouse Expression Set 430 | 1430421_a_at | - | - |
| Mouse Expression Set 430 | 1430421_PM_a_at | - | - |
| Murine Genome U74v2 Set | 95442_PM_at | - | - |
| Murine Genome U74v2 Set | 95442_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430421_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430421_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430421_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430421_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 922946 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4670594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004670594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257823 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2601378 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2601378 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257823 | - | - |
| mouse-6 v1.1 (Illumina) | scl0235043.1_144-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003600.1_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4670594 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003600.1_3-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0235043.1_144-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2601379 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2601378 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2601379 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2601378 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003600.1_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0235043.1_144-S | - | - |

Tmem205
Genomic Map