Assay Details
| Gene Symbol: Necab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178617.4 | NP_848732.3 | 6 - 7 | 708 | 87 | |
| GenBank mRNA | AK039240.1 | - | 6 - 7 | 524 | 87 | |
| AK050307.1 | - | 7 - 8 | 605 | 87 | ||
| AK081951.1 | - | 6 - 7 | 600 | 87 | ||
| AY278200.2 | - | 6 - 7 | 691 | 87 | ||
| BC067055.1 | - | 6 - 7 | 708 | 30545678 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Necab1
Genomic Map