Assay Details
| Gene Symbol: Inpp5f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178641.5 | NP_848756.2 | 14 - 15 | 1921 | 104 | |
| GenBank mRNA | AK030870.1 | - | 14 - 15 | 1923 | 104 | |
| AK129249.1 | - | 14 - 15 | 1878 | 104 | ||
| AK154996.1 | - | 14 - 15 | 1926 | 104 | ||
| AK217446.1 | - | 3 - 4 | 294 | 104 | ||
| BC050127.1 | - | 2 - 3 | 282 | 104 | ||
| BC125437.1 | - | 14 - 15 | 1809 | 40129952 | 104 | |
| BC137700.1 | - | 14 - 15 | 1809 | 8860719 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5f
Genomic Map