Assay Details
| Gene Symbol: Inpp5f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178641.5 | NP_848756.2 | 3 - 4 | 542 | 127 | |
| GenBank mRNA | AK030870.1 | - | 3 - 4 | 544 | 127 | |
| AK039468.1 | - | 2 - 3 | 540 | 127 | ||
| AK129249.1 | - | 3 - 4 | 499 | 127 | ||
| AK150418.1 | - | 3 - 4 | 582 | 127 | ||
| AK154996.1 | - | 3 - 4 | 547 | 127 | ||
| AK211511.1 | - | 2 - 3 | 218 | 127 | ||
| AK218585.1 | - | 2 - 3 | 217 | 127 | ||
| BC125437.1 | - | 3 - 4 | 430 | 40129952 | 127 | |
| BC137700.1 | - | 3 - 4 | 430 | 8860719 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5f
Genomic Map