Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200000242.mm.1 |
NM_001190400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000242.mm.1 |
NM_001190400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000242.mm.1 |
NM_178663.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000242.mm.1 |
NM_178663.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000242.mm.1 |
TC0200000033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000241.mm.1 |
NM_001190400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000241.mm.1 |
NM_001190400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000241.mm.1 |
NM_178663.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000241.mm.1 |
NM_178663.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000241.mm.1 |
TC0200000033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000238.mm.1 |
NM_001190400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000238.mm.1 |
NM_001190400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000238.mm.1 |
NM_178663.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000238.mm.1 |
NM_178663.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000238.mm.1 |
TC0200000033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000243.mm.1 |
NM_001190400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000243.mm.1 |
NM_001190400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000243.mm.1 |
NM_178663.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000243.mm.1 |
NM_178663.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000243.mm.1 |
TC0200000033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000240.mm.1 |
NM_001190400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000240.mm.1 |
NM_001190400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000240.mm.1 |
NM_178663.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000240.mm.1 |
NM_178663.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000240.mm.1 |
TC0200000033.mm.1 |
MTA TC |