Assay Details
Gene Symbol: Prrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178695.5 | NP_848810.3 | 1 - 2 | 309 | 118 | |
GenBank mRNA | AK035530.1 | - | 1 - 2 | 134 | 118 | |
AK037087.1 | - | 1 - 2 | 229 | 118 | ||
AK038703.1 | - | 1 - 2 | 309 | 118 | ||
BC099390.1 | - | 1 - 2 | 226 | 30715383 | 118 | |
BC151011.1 | - | 1 - 2 | 239 | 9087924 | 118 | |
BC151019.1 | - | 1 - 2 | 239 | 9087932 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK035530.1, AK037087.1, AK038703.1, BC099390.1, BC151011.1, BC151019.1, NM_178695.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
