Assay Details
| Gene Symbol: Prrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178695.5 | NP_848810.3 | 2 - 3 | 428 | 76 | |
| GenBank mRNA | AK035530.1 | - | 2 - 3 | 253 | 76 | |
| AK037087.1 | - | 2 - 3 | 348 | 76 | ||
| AK038703.1 | - | 2 - 3 | 428 | 76 | ||
| AK166405.1 | - | 1 - 2 | 389 | 76 | ||
| BC099390.1 | - | 2 - 3 | 345 | 30715383 | 76 | |
| BC115470.2 | - | 2 - 3 | 174 | 40100436 | 76 | |
| BC151011.1 | - | 2 - 3 | 358 | 9087924 | 76 | |
| BC151019.1 | - | 2 - 3 | 358 | 9087932 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prrg4
Genomic Map