Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900017225.mm.1 |
NM_001199556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017225.mm.1 |
NM_001199556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017225.mm.1 |
NM_178737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017225.mm.1 |
NM_178737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017225.mm.1 |
TC0900002083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017227.mm.1 |
NM_001199556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017227.mm.1 |
NM_001199556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017227.mm.1 |
NM_178737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017227.mm.1 |
NM_178737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017227.mm.1 |
TC0900002083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017224.mm.1 |
NM_001199556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017224.mm.1 |
NM_001199556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017224.mm.1 |
NM_178737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017224.mm.1 |
NM_178737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017224.mm.1 |
TC0900002083.mm.1 |
MTA TC |