Assay Details
| Gene Symbol: Tm6sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293795.1 | NP_001280724.1 | 1 - 2 | 170 | 91 | |
| NM_181540.4 | NP_853518.1 | 1 - 2 | 170 | 91 | ||
| GenBank mRNA | AK077126.1 | - | 1 - 2 | 163 | 91 | |
| BB869951.1 | - | 1 - 2 | 171 | 91 | ||
| BC024498.1 | - | 1 - 2 | 158 | 4982620 | 91 | |
More Information
Set Membership: |
|

Tm6sf2
Genomic Map