Assay Details
Gene Symbol: Cmtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY241873.1 | - | 803 | 80 | ||
AY241874.1 | - | 1237 | 80 | |||
AY369078.2 | - | 698 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972464 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078935 | AY369078.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078935 | AY241874.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106376 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117489 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158594 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972464 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078935 | AY369078.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078935 | AY241874.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY369078.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY241873.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY241874.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY162281.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY172740.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127839 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127853 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623672 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623710 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623661 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158594 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01816083 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623758 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01816102 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117489 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106376 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623762 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623736 | - | - |
Whole Mouse Genome G4122A | A_52_P139488 | AY369078.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139488 | AY241873.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139488 | AY241874.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139488 | AY162281.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139488 | AY172740.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106376 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06623672 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117489 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158594 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06623762 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06623710 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06756334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127817 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06883320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06937440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127827 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10581058 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581054 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581059 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581052 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431710_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10581049 | - | - |
Mouse Expression Set 430 | 1431710_at | - | - |
Mouse Expression Set 430 | 1431710_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431710_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431710_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 719400 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2260494 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2260494 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002260494 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706847 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2706847 | - | - |
mouse-6 v1.1 (Illumina) | scl0353509.1_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2260494 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0353509.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260494 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2809355 | - | - |
