Assay Details
| Gene Symbol: Cptp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024472.4 | NP_077792.2 | 3 | 2148 | 82 | |
| GenBank mRNA | AK040664.1 | - | 3 | 2149 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441153 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084522 | NM_024472.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084522 | AK040664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980119 | NM_024472.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980119 | AK040664.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630240 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592374 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149158 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980119 | NM_024472.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980119 | AK040664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084522 | NM_024472.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084522 | AK040664.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441127 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149158 | - | - |
| Whole Mouse Genome G4122A | A_51_P211488 | AK089229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P211488 | BC002216.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105744 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149158 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03239345 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03239350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03239356 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03239344 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03861235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03239354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03861274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03861251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03239345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03861256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441150 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10519221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519222 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448693_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10519219 | - | - |
| Mouse Genome 430 2.0 Array | 1448693_at | - | - |
| Mouse Genome 430 2.0 Array | 1448693_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 79554_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 79554_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400034910.mm.1 | TC0400004158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400034910.mm.1 | NM_024472.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400034910.mm.1 | NM_024472.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400034913.mm.1 | TC0400004158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400034913.mm.1 | NM_024472.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400034913.mm.1 | NM_024472.4 | Refseq |
| Mouse Expression Set 430 | 1448693_at | - | - |
| Mouse Expression Set 430 | 1448693_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448693_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448693_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448693_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448693_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 717303 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4200537 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106400035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200537 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250122 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250122 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251114 | - | - |
| mouse-6 v1.1 (Illumina) | scl23426.4_50-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200537 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106400035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23426.4_50-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106400035 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200537 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2939377 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251114 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2939377 | - | - |

Cptp
Genomic Map