Assay Details
| Gene Symbol: Sike1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025679.3 | NP_079955.1 | 5 | 1948 | 67 | |
| GenBank mRNA | AK033553.1 | - | 5 | 1952 | 67 | |
| AK035590.1 | - | 5 | 1962 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383780 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383782 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383779 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383781 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P135357 | AK033553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P135357 | AK035590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P135357 | AK017694.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P135357 | NM_025679.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115192 | BC004090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115192 | AK033553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115192 | AK035590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115192 | AK014113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115192 | AK014247.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234008 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171944 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P135357 | AK033553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P135357 | AK035590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P135357 | AK017694.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P135357 | NM_025679.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115192 | BC004090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115192 | AK033553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115192 | AK035590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115192 | AK014113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115192 | AK014247.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22383768 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05012545 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171944 | - | - |
| Whole Mouse Genome G4122A | A_52_P484799 | AK033553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484799 | AK035590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P484799 | NM_025679.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131260 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171944 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02773136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02773133 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02758114 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02773134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383782 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383781 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383779 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22383780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22383827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02733441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02774282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02532224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02875605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02761447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03172065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02761448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02733443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02678698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03090993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02593567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02761450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02772079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03172071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03180395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02630051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22383791 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494837 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438442_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450679_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422595_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422296_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422594_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443226_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494832 | - | - |
| Mouse Genome 430 2.0 Array | 1450679_at | - | - |
| Mouse Genome 430 2.0 Array | 1422296_at | - | - |
| Mouse Genome 430 2.0 Array | 1422594_at | - | - |
| Mouse Genome 430 2.0 Array | 1450679_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443226_at | - | - |
| Mouse Genome 430 2.0 Array | 1422595_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422595_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422594_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422296_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443226_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66641_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66641_at | - | - |
| Mouse Expression Set 430 | 1422595_s_at | - | - |
| Mouse Expression Set 430 | 1422296_at | - | - |
| Mouse Expression Set 430 | 1422594_PM_at | - | - |
| Mouse Expression Set 430 | 1422296_PM_at | - | - |
| Mouse Expression Set 430 | 1450679_at | - | - |
| Mouse Expression Set 430 | 1422595_PM_s_at | - | - |
| Mouse Expression Set 430 | 1422594_at | - | - |
| Mouse Expression Set 430 | 1450679_PM_at | - | - |
| Mouse Expression Set 430 | 1443226_at | - | - |
| Mouse Expression Set 430 | 1443226_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422296_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422594_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450679_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450679_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422296_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422595_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422595_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422594_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422595_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422594_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422595_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450679_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422296_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422296_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450679_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422594_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443226_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438442_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443226_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438442_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 305197 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2760064 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2760064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670897 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670897 | - | - |
| mouse-6 v1.1 (Illumina) | scl22798.5_507-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22798.5_507-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2670897 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2670897 | - | - |

Sike1
Genomic Map