Assay Details
Gene Symbol: Ms4a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027209.3 | NP_081485.2 | 7 | 1375 | 140 | |
GenBank mRNA | AF237909.1 | - | 7 | 1375 | 140 | |
AK075671.1 | - | 7 | 1352 | 140 | ||
AK149773.1 | - | 409 | 140 | |||
AK172345.1 | - | 3123 | 140 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180176 | AK172345.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180176 | NM_027209.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180176 | AF237909.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180176 | AK149773.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134040 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337837 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290480 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454212 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145165 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420320 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307693 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99256 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171324 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171864 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180176 | AK172345.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180176 | NM_027209.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180176 | AF237909.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180176 | AK149773.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422300 | AK152258.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422300 | AK150397.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422300 | NM_028595.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422300 | AK149773.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422300 | AK172345.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956485 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956506 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956495 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956487 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956511 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956488 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31956508 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171864 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171324 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09106589 | - | - |
Whole Mouse Genome G4122A | A_51_P422300 | AK152258.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P422300 | AK150397.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P422300 | NM_028595.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P422300 | AK149773.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P422300 | AK172345.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115991 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171324 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171864 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14207230 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14207215 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14309775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31956562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14476665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956465 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14208729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956463 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956527 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956495 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956528 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956540 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956484 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14443739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956541 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14207213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956545 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956537 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956485 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956507 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14372147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956487 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956555 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956535 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956522 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14207226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956503 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14385071 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14443743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14443732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956481 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956490 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956511 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956486 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956506 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31956552 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10461626 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461628 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461623 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461627 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461625 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461624 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1439956_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418826_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10461622 | - | - |
Mouse Genome 430 2.0 Array | 1442219_at | - | - |
Mouse Genome 430 2.0 Array | 1442219_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1439956_at | - | - |
Mouse Genome 430 2.0 Array | 1418826_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418826_at | - | - |
Mouse Genome 430 2.0 Array | 1439956_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69774_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69774_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900002775.mm.1 | NM_027209.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002775.mm.1 | NM_027209.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002775.mm.1 | TC1900000225.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418826_PM_at | - | - |
Mouse Expression Set 430 | 1418826_at | - | - |
Mouse Expression Set 430 | 1439956_at | - | - |
Mouse Expression Set 430 | 1439956_PM_at | - | - |
Murine Genome U74v2 Set | 97322_PM_at | - | - |
Murine Genome U74v2 Set | 97322_at | - | - |
Mouse Genome 430A 2.0 Array | 1418826_at | - | - |
Mouse Genome 430A 2.0 Array | 1418826_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418826_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418826_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4920279 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004920279 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254692 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254692 | - | - |
mouse-6 v1.1 (Illumina) | scl53360.9.1_13-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4920279 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl53360.9.1_13-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920279 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1254692 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254692 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53360.9.1_13-S | - | - |
