Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900002765.mm.1 |
NM_027209.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002765.mm.1 |
NM_027209.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002765.mm.1 |
TC1900000225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002763.mm.1 |
NM_027209.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002763.mm.1 |
NM_027209.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002763.mm.1 |
TC1900000225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002773.mm.1 |
NM_027209.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002773.mm.1 |
NM_027209.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002773.mm.1 |
TC1900000225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002769.mm.1 |
NM_027209.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002769.mm.1 |
NM_027209.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002769.mm.1 |
TC1900000225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002767.mm.1 |
NM_027209.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002767.mm.1 |
NM_027209.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002767.mm.1 |
TC1900000225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002764.mm.1 |
NM_027209.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002764.mm.1 |
NM_027209.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002764.mm.1 |
TC1900000225.mm.1 |
MTA TC |