Assay Details
| Gene Symbol: Slc25a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133668.3 | NP_598429.1 | 5 | 552 | 65 | |
| GenBank mRNA | AK077274.1 | - | 5 | 553 | 65 | |
| AK077723.1 | - | 5 | 609 | 65 | ||
| AK088013.1 | - | 5 | 557 | 65 | ||
| AK150641.1 | - | 5 | 557 | 65 | ||
| AK150729.1 | - | 5 | 557 | 65 | ||
| AK150870.1 | - | 5 | 596 | 65 | ||
| AK151088.1 | - | 5 | 557 | 65 | ||
| AK151518.1 | - | 5 | 557 | 65 | ||
| AK151887.1 | - | 5 | 557 | 65 | ||
| AK152006.1 | - | 5 | 557 | 65 | ||
| AK152292.1 | - | 5 | 557 | 65 | ||
| AK152341.1 | - | 5 | 553 | 65 | ||
| AK152713.1 | - | 5 | 556 | 65 | ||
| AK167197.1 | - | 5 | 557 | 65 | ||
| AK168129.1 | - | 5 | 598 | 65 | ||
| AK168580.1 | - | 5 | 553 | 65 | ||
| AK188364.1 | - | 2 | 236 | 65 | ||
| AK195219.1 | - | 2 | 187 | 65 | ||
| AK212727.1 | - | 2 | 220 | 65 | ||
| AK216642.1 | - | 2 | 220 | 65 | ||
| BC004030.1 | - | 5 | 532 | 65 | ||
| BC018161.1 | - | 5 | 567 | 2650436 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340133 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042071 | BC004030.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042071 | BC018161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042071 | AK200877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042071 | AK202780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042071 | AK200546.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050448 | AK037421.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050448 | NM_133668.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050448 | AK077274.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666282 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157928 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101479 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050448 | AK037421.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050448 | NM_133668.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050448 | AK077274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042071 | BC004030.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042071 | BC018161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042071 | AK200877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042071 | AK202780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042071 | AK200546.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269652 | BC004030.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269652 | BC018161.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269652 | AK216642.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269652 | AK195219.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269652 | AK152341.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27340131 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101479 | - | - |
| Whole Mouse Genome G4122A | A_51_P269652 | BC004030.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269652 | BC018161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269652 | AK216642.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269652 | AK195219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269652 | AK152341.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124886 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101479 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08309086 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08309075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08309072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08309087 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08309069 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27340139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27340087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08870226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08262394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08309099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08272109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08309075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08413129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08309070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08309087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08413128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08276778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27340147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08272107 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10371879 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371880 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371884 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440302_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416300_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10371877 | - | - |
| Mouse Genome 430 2.0 Array | 1416300_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416300_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1440302_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440302_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18674_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18674_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000020377.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020377.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020377.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020370.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020370.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020370.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020374.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020374.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020374.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020378.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020378.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020378.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020369.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020369.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020369.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020392.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020392.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020392.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020375.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020375.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020375.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020384.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020384.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020384.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020366.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020366.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020366.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020372.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020372.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020372.mm.1 | TC1000002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000020390.mm.1 | NM_133668.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020390.mm.1 | NM_133668.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000020390.mm.1 | TC1000002668.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416300_a_at | - | - |
| Mouse Expression Set 430 | 1416300_PM_a_at | - | - |
| Mouse Expression Set 430 | 1440302_at | - | - |
| Mouse Expression Set 430 | 1440302_PM_at | - | - |
| Murine Genome U74v2 Set | 93506_PM_at | - | - |
| Murine Genome U74v2 Set | 93506_at | - | - |
| Murine Genome U74v2 Set | 136620_PM_r_at | - | - |
| Murine Genome U74v2 Set | 136620_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416300_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416300_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416300_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416300_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440302_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440302_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 593450 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 670112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2751046 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2751046 | - | - |
| mouse-6 v1.1 (Illumina) | scl37617.5_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670112 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37617.5_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2751046 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2960467 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2751046 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2960467 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37617.5_6-S | - | - |

Slc25a3
Genomic Map