Assay Details
| Gene Symbol: Vmn1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134187.3 | NP_598948.1 | 1 | 944 | 144 | |
| GenBank mRNA | AF394951.1 | - | 1 | 944 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005165 | AF394951.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005165 | NM_134187.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163985 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005165 | AF394951.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005165 | NM_134187.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422465 | NM_001166710.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422465 | AF394951.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422465 | NM_134187.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24477851 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163985 | - | - |
| Whole Mouse Genome G4122A | A_51_P422465 | NM_001166710.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422465 | AF394951.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422465 | NM_134187.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102968 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125371 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05157236 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05157239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24477840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05256820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05123666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05483333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05709138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05745856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24477819 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10538570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544994 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10538569 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10544993 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10538567 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600005287.mm.1 | TC0600000683.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600005287.mm.1 | NM_134187.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600005287.mm.1 | NM_134187.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 681003 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101740500 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1850048 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101740500 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1850048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2483806 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2483806 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238552 | - | - |
| mouse-6 v1.1 (Illumina) | scl29995.1.1_130-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_31982576-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101740500 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29995.1.1_130-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850048 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101740500 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2483806 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2483806 | - | - |

Vmn1r8
Genomic Map