Assay Details
| Gene Symbol: B3gnt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036740.2 | NP_001031817.1 | 2 | 1377 | 105 | |
| NM_146184.4 | NP_666296.1 | 1 | 1769 | 105 | ||
| GenBank mRNA | AK042388.1 | - | 1 | 1752 | 105 | |
| AK154220.1 | - | 1364 | 105 | |||
| AK154901.1 | - | 2 | 1333 | 105 | ||
| BC025206.1 | - | 2 | 1363 | 5037295 | 105 | |
| BY554292.1 | - | 1 | 437 | 105 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
poly-N-acetyllactosamine biosynthetic process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
protein N-acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
protein glycosylation
poly-N-acetyllactosamine biosynthetic process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
protein N-acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010985 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010987 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010988 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010986 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010984 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | NM_146184.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | AK042388.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | AK154220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | AK154901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | BC025206.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | NM_146184.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | AK042388.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | AK154220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | AK154901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170178 | BC025206.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110690 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167766 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | NM_146184.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | AK042388.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | AK154220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | AK154901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | BC025206.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | NM_146184.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | AK042388.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | AK154220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | AK154901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170178 | BC025206.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25011000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010999 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167766 | - | - |
| Whole Mouse Genome G4122A | A_51_P170178 | NM_146184.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | AK042388.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | AK154220.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | AK154901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | BC025206.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | NM_146184.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | AK042388.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | AK154220.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | AK154901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170178 | BC025206.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110976 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167766 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06400185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06400180 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06614029 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06400178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06400175 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06400179 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010984 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010985 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010988 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010986 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06400178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06614022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25011002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010980 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551170 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425128_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551169 | - | - |
| Mouse Genome 430 2.0 Array | 1425128_at | - | - |
| Mouse Genome 430 2.0 Array | 1425128_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232984_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232984_at | - | - |
| Mouse Expression Set 430 | 1425128_at | - | - |
| Mouse Expression Set 430 | 1425128_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425128_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425128_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425128_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425128_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 433761 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5360372 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360372 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2614889 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2614889 | - | - |
| mouse-6 v1.1 (Illumina) | scl32924.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360372 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32924.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360372 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3149776 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2614889 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2614889 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3149776 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32924.1_1-S | - | - |

B3gnt8
Genomic Map