Assay Details
| Gene Symbol: Olfr1309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146447.1 | NP_666658.1 | 1 | 914 | 142 | |
| GenBank mRNA | BC128010.1 | - | 1 | 918 | 40136004 | 142 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886491 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154885 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154885 | - | - |
| Whole Mouse Genome G4122A | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195735 | NM_146447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195735 | BC128010.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123533 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21526915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01520877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01162895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01419505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02074381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02145197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01728786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21526941 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10485803 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10485802 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200036240.mm.1 | NM_146447.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036240.mm.1 | NM_146447.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036240.mm.1 | TC0200004337.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6450288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006450288 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2611973 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2611973 | - | - |
| mouse-6 v1.1 (Illumina) | scl18873.2.10_226-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6450288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18873.2.10_226-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6450288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2961541 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2961541 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18873.2.10_226-S | - | - |

Olfr1309
Genomic Map