Assay Details
Gene Symbol: Olfr108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146465.2 | NP_666676.1 | 1 | 91 | 88 | |
GenBank mRNA | BC132008.1 | - | 1 | 91 | 40130285 | 88 |
BC132012.1 | - | 1 | 91 | 40130289 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079791 | BC132008.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079791 | BC132012.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079791 | NM_146465.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129549 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079791 | BC132008.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079791 | BC132012.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079791 | NM_146465.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P459308 | BC132008.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P459308 | BC132012.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P459308 | NM_146465.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168735 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129549 | - | - |
Whole Mouse Genome G4122A | A_51_P459308 | BC132008.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P459308 | BC132012.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P459308 | NM_146465.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124889 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114861 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129549 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13075917 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13075909 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13075921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168753 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13075917 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168758 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168738 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168753 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168747 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168755 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168728 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168730 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13536614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168760 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168740 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10445136 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10445135 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6840168 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6840168 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006840168 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600380 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2600380 | - | - |
mouse-6 v1.1 (Illumina) | scl8519.1.1_49-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6840168 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl8519.1.1_49-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840168 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2898062 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2600380 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2898062 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600380 | - | - |
