Assay Details
Gene Symbol: Olfr183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146485.2 | NP_666696.2 | 2 | 519 | 70 | |
GenBank mRNA | DR065568.1 | - | 2 | 538 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073445 | NM_146485.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54956 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100817 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169213 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073445 | NM_146485.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P334499 | NM_146485.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P334499 | DR065568.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P334499 | DR065627.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793874 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793917 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793926 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793922 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793897 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793916 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793929 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30793908 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100817 | - | - |
Whole Mouse Genome G4122A | A_52_P334499 | NM_146485.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P334499 | DR065568.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P334499 | DR065627.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137663 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100817 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12864586 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12864581 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12864583 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12864571 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12864576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30793923 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793889 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793849 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793894 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793897 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793916 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793911 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12864592 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793933 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793927 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12944872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12891852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12796690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793930 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793922 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793860 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12771911 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793926 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793923 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13050572 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793931 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793917 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793907 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793925 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793864 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13054075 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12756251 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30793883 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10436410 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10436411 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10436409 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 597019 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3800538 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003800538 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2747771 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2747771 | - | - |
mouse-6 v1.1 (Illumina) | scl49009.2.89_75-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3800538 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl49009.2.89_75-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800538 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2747771 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2747771 | - | - |
