Assay Details
| Gene Symbol: Olfr96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146514.1 | NP_666725.1 | 1 | 252 | 114 | |
| GenBank mRNA | BC120652.1 | - | 1 | 288 | 40129575 | 114 | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Ambion siRNA available  Amplicon length greater than or equal to 101  G-protein coupled receptor  Mammalian Gene Collection  Detect Genomic DNA  Recommended Assay  Mus musculus  1700  Inventoried  Both primers and probe map within a single exon | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
					
						
							Agilent
					
						
							Affymetrix
					
						
							ABI
					
						
							Illumina
					
					
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119214 | NM_146514.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119214 | BC120652.1 | mouse_transcriptome | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122775 | - | - | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119214 | NM_146514.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119214 | BC120652.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P211112 | NM_146514.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P211112 | BC120652.1 | mouse_transcriptome | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167923 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167917 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167948 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167922 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167928 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167927 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167946 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167920 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167932 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167925 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167933 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167947 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167915 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167936 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167919 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167931 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167929 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167935 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167916 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31167943 | - | - | 
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122775 | - | - | 
| Whole Mouse Genome G4122A | A_52_P211112 | NM_146514.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P211112 | BC120652.1 | mouse_transcriptome | 
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118447 | - | - | 
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100043 | - | - | 
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122775 | - | - | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13074633 | - | - | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13074626 | - | - | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13074636 | - | - | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13074637 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167920 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167917 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167929 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167932 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167981 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167948 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167886 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167963 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167889 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167947 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167912 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167906 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167923 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167897 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167925 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167892 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167913 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167914 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167983 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167930 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167898 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167974 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167960 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167894 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167883 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167935 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167969 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167928 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167911 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167922 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167965 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167909 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167908 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167972 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167902 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167901 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31167916 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167932 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302216 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167902 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167912 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167942 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167917 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167901 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167897 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302212 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167914 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167913 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167947 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167929 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167946 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167936 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167935 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167943 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167920 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167908 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167927 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167933 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13354842 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167931 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167906 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167928 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167909 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13538239 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13489081 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167911 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167916 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167930 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167925 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167900 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302217 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31167923 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445118 | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10445117 | - | - | 
| Affymetrix Mouse Clariom S Array | PSR1700007679.mm.1 | TC1700000762.mm.1 | MTA TC | 
| Affymetrix Mouse Clariom S Array | PSR1700007681.mm.1 | TC1700000762.mm.1 | MTA TC | 
| Affymetrix Mouse Clariom S Array | PSR1700007680.mm.1 | NM_146514.1.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR1700007680.mm.1 | NM_146514.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR1700007680.mm.1 | TC1700000762.mm.1 | MTA TC | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 689405 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2630129 | - | - | 
| Illumina mouse-6 v1.0 expression beadchip | 2630129 | - | - | 
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630129 | - | - | 
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2722883 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2722883 | - | - | 
| mouse-6 v1.1 (Illumina) | scl50735.1.1_119-S | - | - | 
| Illumina Sentrix mouse-6.v1 bead array | 2630129 | - | - | 
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50735.1.1_119-S | - | - | 
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630129 | - | - | 
| MouseRef-8 v2 Expression BeadChip | ILMN_2722883 | - | - | 
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2722883 | - | - | 
 Back To Top
 Back To Top
 Olfr96
Olfr96 Genomic Map
Genomic Map Back To Top
 Back To Top