Assay Details
Gene Symbol: Olfr365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146662.1 | NP_666873.1 | 1 | 457 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081675 | NM_146662.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081675 | NM_146662.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426708 | NM_146662.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141249 | - | - |
Whole Mouse Genome G4122A | A_51_P426708 | NM_146662.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124627 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02085334 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02085335 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02085332 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02085329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141273 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01894569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21141277 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10471775 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10471777 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10471776 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10471774 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 530866 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6650563 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6650563 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006650563 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624376 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2624376 | - | - |
mouse-6 v1.1 (Illumina) | scl0258656.1_328-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6650563 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258656.1_328-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650563 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2624376 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2624376 | - | - |
