Assay Details
Gene Symbol: Olfr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146682.1 | NP_666893.1 | 1 | 853 | 122 | |
GenBank mRNA | BC138072.1 | - | 1 | 879 | 8861091 | 122 |
BC138073.1 | - | 1 | 879 | 8861092 | 122 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119204 | BC138073.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119204 | NM_146682.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119204 | BC138072.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92679 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170349 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119204 | BC138073.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119204 | NM_146682.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119204 | BC138072.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204982 | BC138073.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204982 | Y15525.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204982 | NM_146682.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204982 | BC138072.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959641 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959639 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959608 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170349 | - | - |
Whole Mouse Genome G4122A | A_51_P204982 | BC138073.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204982 | Y15525.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204982 | NM_146682.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P204982 | BC138072.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110844 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170349 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14339599 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14392523 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14339598 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14392514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14392526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14392523 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14445345 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14490350 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14178406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14496830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14445346 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14490344 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959615 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959634 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959635 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959633 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14490343 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14490353 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14397812 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10466291 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10466290 | - | - |
Affymetrix Mouse Clariom S Array | PSR1900010774.mm.1 | NM_146682.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900010774.mm.1 | NM_146682.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900010774.mm.1 | TC1900001156.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900010775.mm.1 | TC1900001156.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900010773.mm.1 | TC1900001156.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 636219 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4670609 | - | - |
MouseWG-6 v2 Expression BeadChip | 4570280 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4570280 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4670609 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004570280 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004670609 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224313 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213856 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213856 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224313 | - | - |
mouse-6 v1.1 (Illumina) | scl52710.2.242_50-S | - | - |
mouse-6 v1.1 (Illumina) | scl0140479.2_128-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4670609 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4570280 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52710.2.242_50-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0140479.2_128-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670609 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570280 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1213856 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1224313 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224313 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213856 | - | - |
