Assay Details
| Gene Symbol: Mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172252.3 | NP_758456.2 | 7 | 950 | 83 | |
| GenBank mRNA | AK020739.1 | - | 7 | 951 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969231 | AK020739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969231 | NM_172252.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132322 | AK003285.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132322 | BC028768.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214201 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123985 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106613 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150229 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132322 | AK003285.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132322 | BC028768.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969231 | AK020739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969231 | NM_172252.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423625 | AK003285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423625 | AK012784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423625 | AK020739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423625 | BC028768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423625 | NM_172252.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104843 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132322 | AK003285.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132322 | BC028768.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14305584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14268321 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14305577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460156 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460153 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452580_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456275_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460151 | - | - |
| Mouse Genome 430 2.0 Array | 1437795_at | - | - |
| Mouse Genome 430 2.0 Array | 1452580_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437795_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456275_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452580_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456275_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 353242_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 353242_at | - | - |
| Mouse Expression Set 430 | 1452580_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452580_a_at | - | - |
| Mouse Expression Set 430 | 1456275_PM_at | - | - |
| Mouse Expression Set 430 | 1456275_at | - | - |
| Murine Genome U74v2 Set | 101097_at | - | - |
| Murine Genome U74v2 Set | 101097_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452580_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456275_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452580_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456275_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452580_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452580_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456275_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456275_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 416107 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1500368 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106660139 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500368 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2640313 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2543346 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2640313 | - | - |
| mouse-6 v1.1 (Illumina) | scl52785.8.1_26-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500368 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106660139 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52785.8.1_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106660139 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500368 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2640313 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2640313 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2543346 | - | - |

Mrpl21
Genomic Map