Assay Details
| Gene Symbol: Cxcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178241.4 | NP_839972.1 | 2 | 999 | 98 | |
| GenBank mRNA | AY227797.1 | - | 1 | 951 | 98 | |
| AY390263.1 | - | 2 | 999 | 98 | ||
| AY749637.1 | - | 1 | 1325 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chemotaxis
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
receptor internalization
interleukin-8-mediated signaling pathway
chemokine-mediated signaling pathway
signal transducer activity
interleukin-8 receptor activity
G-protein coupled receptor activity
chemokine receptor activity
C-X-C chemokine receptor activity
interleukin-8 binding
chemotaxis
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
receptor internalization
interleukin-8-mediated signaling pathway
chemokine-mediated signaling pathway
signal transducer activity
interleukin-8 receptor activity
G-protein coupled receptor activity
chemokine receptor activity
C-X-C chemokine receptor activity
interleukin-8 binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103249 | NM_178241.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522812 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177682 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103249 | NM_178241.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193808 | AY390263.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193808 | AY749637.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193808 | NM_178241.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P193808 | AY227797.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177682 | - | - |
| Whole Mouse Genome G4122A | A_51_P193808 | AY390263.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193808 | AY749637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193808 | NM_178241.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193808 | AY227797.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122214 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20347235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01082882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00564976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00607039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00450119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00835529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00564979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00072949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00219202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00564981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00450107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00607041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01084575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00564973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00450120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01082885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20347290 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10355551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355552 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355553 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10355550 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100020776.mm.1 | NM_178241.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100020776.mm.1 | NM_178241.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100020776.mm.1 | TC0100002580.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100020774.mm.1 | NM_178241.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100020774.mm.1 | NM_178241.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100020774.mm.1 | TC0100002580.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 461580 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1580100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1580100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001580100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588216 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588216 | - | - |
| mouse-6 v1.1 (Illumina) | scl16618.1.1_330-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1580100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl16618.1.1_330-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2588216 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2588216 | - | - |

Cxcr1
Genomic Map