Assay Details
| Gene Symbol: Gjd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178596.2 | NP_848711.2 | 1 | 53 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059763 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059762 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059758 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059760 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059764 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059761 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167032 | NM_178596.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167032 | NM_178596.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P463340 | NM_178596.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28059797 | - | - |
| Whole Mouse Genome G4122A | A_52_P463340 | NM_178596.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113785 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092052 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092061 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09734557 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092055 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092058 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092050 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059762 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059758 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059761 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059760 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059763 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059764 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28059766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28059789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09016021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09056603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09119297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09298952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09734561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09274186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09329752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09056595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09138945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09056596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09119294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09411837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09092052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09056601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09734560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28059771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390706 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390705 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100034010.mm.1 | NM_178596.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034010.mm.1 | NM_178596.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034010.mm.1 | TC1100003692.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7050082 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7050082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2750021 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2750021 | - | - |
| mouse-6 v1.1 (Illumina) | scl39613.1.1_137-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050082 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39613.1.1_137-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2750021 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2750021 | - | - |

Gjd3
Genomic Map